Assay Details
Target Gene Details
Entrez Gene ID: | 407977 |
Gene Name: | TNFSF12-TNFSF13 readthrough |
Gene Aliases: |
TWE-PRIL |
Location: |
Chr.17:7549058-7561608 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFSF12-TNFSF13 | NM_172089.3 | NP_742086.1 | ||
| AY081051.1 | AAL90443.1 |
Target Gene Details
Entrez Gene ID: | 8741 |
Gene Name: | tumor necrosis factor superfamily member 13 |
Gene Aliases: |
APRIL, CD256, TALL-2, TALL2, TNLG7B, TRDL-1, UNQ383/PRO715, ZTNF2 |
Location: |
Chr.17:7558292-7561608 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFSF13 | NM_001198622.1 | 1 | 583 | NP_001185551.1 |
| NM_001198623.1 | 1 | 583 | NP_001185552.1 | |
| NM_001198624.1 | NP_001185553.1 | |||
| NM_003808.3 | 1 | 583 | NP_003799.1 | |
| NM_172087.2 | 1 | 583 | NP_742084.1 | |
| NM_172088.2 | 1 | 583 | NP_742085.1 | |
| NR_073490.2 | 1 | 583 | ||
| AF046888.1 | 1 | 116 | AAC61312.1 | |
| AF114011.1 | 1 | 116 | AAF59828.1 | |
| AF114012.1 | AAF59829.1 | |||
| AF114013.1 | 1 | 88 | AAF59830.1 | |
| AF184972.1 | AAF01321.1 | |||
| AI828515.1 | ||||
| AK090698.1 | 1 | 399 | ||
| AK225556.1 | 1 | 127 | ||
| AK301221.1 | 1 | 116 | ||
| AK314518.1 | 1 | 122 | ||
| AY081050.1 | 1 | 107 | AAL90442.1 | |
| AY358880.1 | AAQ89239.1 | |||
| BC008042.2 | AAH08042.1 | |||
| BI764562.1 | 1 | 583 | ||
| BI910977.1 | 1 | 576 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion |
|
| nsv574322 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map