Assay Details
Target Gene Details
Entrez Gene ID: | 862 |
Gene Name: | RUNX1 translocation partner 1 |
Gene Aliases: |
AML1-MTG8, AML1T1, CBFA2T1, CDR, ETO, MTG8, ZMYND2 |
Location: |
Chr.8:91954967-92103365 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1022473 | Chr.8:91854892 - 94250166 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map