Assay Details
Target Gene Details
Entrez Gene ID: | 85439 |
Gene Name: | stonin 2 |
Gene Aliases: |
STN2, STNB, STNB2 |
Location: |
Chr.14:81260650-81427404 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STON2 | NM_001256430.1 | NP_001243359.1 | ||
| NM_033104.3 | 6 | 2907 | NP_149095.2 | |
| XM_006720279.1 | XP_006720342.1 | |||
| XM_011537240.1 | XP_011535542.1 | |||
| XM_011537241.1 | 6 | 2967 | XP_011535543.1 | |
| XM_011537242.1 | XP_011535544.1 | |||
| AB208948.1 | BAD92185.1 | |||
| AF255309.1 | 2 | 2106 | AAK57558.1 | |
| AF380833.1 | 5 | 2641 | AAK76362.1 | |
| AF449430.1 | 5 | 2799 | AAL47008.1 | |
| AK094799.1 | ||||
| BC069359.1 | 5 | 2779 | AAH69359.1 | |
| BC069389.1 | 5 | 2773 | AAH69389.1 | |
| BC069502.1 | 5 | 2779 | AAH69502.1 | |
| BC069587.1 | 5 | 2780 | AAH69587.1 | |
| BC117493.1 | 5 | 2709 | ||
| BC117495.1 | 5 | 2709 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832841 | Chr.14:81121978 - 81277577 on Build GRCh38 | Gain |
|
| esv3635076 | Chr.14:80715257 - 81672373 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map