Assay Details
Target Gene Details
Entrez Gene ID: | 344752 |
Gene Name: | arylacetamide deacetylase like 2 |
Gene Aliases: |
- |
Location: |
Chr.3:151733916-151758016 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AADACL2 | NM_207365.3 | 2 | 384 | NP_997248.2 |
| XM_011512753.1 | 2 | 412 | XP_011511055.1 | |
| BC065724.1 | 2 | 377 | AAH65724.1 | |
| BX647585.1 | CAI46075.1 | |||
| CU449274.1 | 2 | 206 |
More Information
Set Membership: |
|

Genomic Map