Assay Details
Target Gene Details
Entrez Gene ID: | 949 |
Gene Name: | scavenger receptor class B member 1 |
Gene Aliases: |
CD36L1, CLA-1, CLA1, HDLQTL6, SR-BI, SRB1 |
Location: |
Chr.12:124777628-124863973 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCARB1 | NM_001082959.1 | NP_001076428.1 | ||
| NM_005505.4 | NP_005496.4 | |||
| AB209436.1 | BAD92673.1 | |||
| AF515445.1 | AAQ08185.1 | |||
| AK124112.1 | ||||
| AK302779.1 | ||||
| AK304730.1 | ||||
| BC080647.1 | AAH80647.1 | |||
| BC093732.1 | AAH93732.1 | |||
| BC112037.1 | AAI12038.1 | |||
| BC143319.1 | ||||
| KU177964.1 | ||||
| KU177965.1 | ||||
| Z22555.1 | CAA80277.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560595 | Chr.12:124737381 - 125019207 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map