Assay Details
Target Gene Details
Entrez Gene ID: | 5079 |
Gene Name: | paired box 5 |
Gene Aliases: |
ALL3, BSAP |
Location: |
Chr.9:36833274-37035949 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAX5 | NM_001280547.1 | 9 | 1961 | NP_001267476.1 |
| NM_001280548.1 | 9 | 1976 | NP_001267477.1 | |
| NM_001280549.1 | 8 | 1831 | NP_001267478.1 | |
| NM_001280550.1 | 7 | 1744 | NP_001267479.1 | |
| NM_001280551.1 | 7 | 1708 | NP_001267480.1 | |
| NM_001280552.1 | 8 | 1874 | NP_001267481.1 | |
| NM_001280553.1 | 8 | 1847 | NP_001267482.1 | |
| NM_001280554.1 | 9 | 1934 | NP_001267483.1 | |
| NM_001280555.1 | 8 | 1763 | NP_001267484.1 | |
| NM_001280556.1 | 9 | 1897 | NP_001267485.1 | |
| NM_016734.2 | 10 | 2063 | NP_057953.1 | |
| NR_103999.1 | 9 | 1933 | ||
| NR_104000.1 | 10 | 1990 | ||
| XM_005251481.4 | 10 | 2684 | XP_005251538.1 | |
| XM_011517896.2 | 11 | 3590 | XP_011516198.1 | |
| XM_011517897.2 | 11 | 2745 | XP_011516199.1 | |
| M96944.1 | 10 | 1690 | AAA58397.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6531 | Chr.9:36832805 - 36876608 on Build GRCh38 | Deletion |
|
| nsv471306 | Chr.9:36740960 - 36848924 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map