Assay Details
Target Gene Details
Entrez Gene ID: | 146722 |
Gene Name: | CD300 molecule like family member f |
Gene Aliases: |
CD300f, CLM-1, CLM1, IREM-1, IREM1, IgSF13, LMIR3, NKIR |
Location: |
Chr.17:74694308-74713059 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD300LF | NM_001289082.1 | NP_001276011.1 | ||
| NM_001289083.1 | NP_001276012.1 | |||
| NM_001289084.1 | NP_001276013.1 | |||
| NM_001289085.1 | NP_001276014.1 | |||
| NM_001289086.1 | NP_001276015.1 | |||
| NM_001289087.1 | NP_001276016.1 | |||
| NM_139018.4 | NP_620587.2 | |||
| NR_110298.1 | ||||
| XM_017024212.1 | 1 | 110 | XP_016879701.1 | |
| XM_017024213.1 | 1 | 117 | XP_016879702.1 | |
| XM_017024214.1 | 1 | 116 | XP_016879703.1 | |
| XM_017024215.1 | XP_016879704.1 | |||
| XM_017024216.1 | 1 | 114 | XP_016879705.1 | |
| XM_017024217.1 | 1 | 112 | XP_016879706.1 | |
| AK092757.1 | BAC03966.1 | |||
| AK298752.1 | ||||
| AK315165.1 | ||||
| AY358545.1 | AAQ88909.1 | |||
| BC028199.1 | AAH28199.1 | |||
| BM147210.1 | ||||
| BP226373.1 | ||||
| DA673169.1 |
Target Gene Details
Entrez Gene ID: | 326624 |
Gene Name: | RAB37, member RAS oncogene family |
Gene Aliases: |
- |
Location: |
Chr.17:74671117-74747335 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB37 | NM_175738.4 | NP_783865.1 | ||
| AK054846.1 | ||||
| AK127073.1 | ||||
| AK290202.1 | ||||
| BC040547.1 | AAH40547.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3282n100 | Chr.17:73814343 - 74719597 on Build GRCh38 | Gain |
|
| esv33995 | Chr.17:74261053 - 74732093 on Build GRCh38 | Loss |
|
| esv2751770 | Chr.17:73837862 - 74716223 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map