Assay Details
Target Gene Details
Entrez Gene ID: | 10952 |
Gene Name: | Sec61 translocon beta subunit |
Gene Aliases: |
- |
Location: |
Chr.9:99222288-99230619 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEC61B | NM_006808.2 | NP_006799.1 | ||
| AJ293399.1 | ||||
| AK223384.1 | BAD97104.1 | |||
| BC001734.1 | AAH01734.1 | |||
| CR456883.1 | CAG33164.1 | |||
| CR542159.1 | CAG46956.1 | |||
| L25085.1 | AAA19706.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621214 | Chr.9:98651384 - 99637088 on Build GRCh38 | Gain |
|
| nsv831666 | Chr.9:99112703 - 99256408 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map