Assay Details
Target Gene Details
Entrez Gene ID: | 1143 |
Gene Name: | cholinergic receptor nicotinic beta 4 subunit |
Gene Aliases: |
- |
Location: |
Chr.15:78623282-78655586 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHRNB4 | NM_000750.3 | NP_000741.1 | ||
| NM_001256567.1 | NP_001243496.1 | |||
| XM_011521186.2 | XP_011519488.1 | |||
| XM_011521187.2 | XP_011519489.1 | |||
| XM_011521191.2 | XP_011519493.1 | |||
| XM_011521192.2 | XP_011519494.1 | |||
| XM_017021887.1 | XP_016877376.1 | |||
| XM_017021888.1 | XP_016877377.1 | |||
| XM_017021889.1 | XP_016877378.1 | |||
| AK310443.1 | ||||
| BC096080.1 | AAH96080.1 | |||
| BC096081.1 | ||||
| BC096082.1 | AAH96082.1 | |||
| BC096083.1 | ||||
| U48861.1 | AAA92123.1 | |||
| U62439.1 | AAB40117.1 | |||
| Y08416.1 | CAA69693.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1628 | Chr.15:78617170 - 78649604 on Build GRCh38 | Insertion |
|
| nsv1045827 | Chr.15:78615314 - 78712300 on Build GRCh38 | Gain |
|
| nsv570202 | Chr.15:78622903 - 78661443 on Build GRCh38 | Loss |
|
| nsv457208 | Chr.15:78615690 - 78727268 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map