Assay Details
Target Gene Details
Entrez Gene ID: | 25792 |
Gene Name: | CDKN1A interacting zinc finger protein 1 |
Gene Aliases: |
LSFR1, NP94, ZNF356 |
Location: |
Chr.9:128166065-128204383 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CIZ1 | NM_001131015.1 | NP_001124487.1 | ||
| NM_012127.2 | NP_036259.2 | |||
| AK023978.1 | BAB14750.1 |
Target Gene Details
Entrez Gene ID: | 1759 |
Gene Name: | dynamin 1 |
Gene Aliases: |
DNM, EIEE31 |
Location: |
Chr.9:128203355-128255248 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
More Information
Set Membership: |
|

Genomic Map