Assay Details
Target Gene Details
Entrez Gene ID: | 149998 |
Gene Name: | lipase I |
Gene Aliases: |
CT17, LPDL, PLA1C, PRED5, mPA-PLA1 beta |
Location: |
Chr.21:14108813-14210953 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIPI | NM_001302998.1 | 9 | 1255 | NP_001289927.1 |
| NM_001302999.1 | 8 | 1165 | NP_001289928.1 | |
| NM_001303000.1 | 9 | 1237 | NP_001289929.1 | |
| NM_001303001.1 | NP_001289930.1 | |||
| NM_198996.3 | 9 | 1298 | NP_945347.2 | |
| AI017218.1 | ||||
| AY197607.1 | 9 | 1282 | AAP37476.1 | |
| JN387910.1 | 9 | 1255 | ||
| JN387911.1 | 10 | 1366 | ||
| JN387912.1 | 7 | 760 | ||
| JN387913.1 | 8 | 1165 | ||
| JN387914.1 | 9 | 1150 | ||
| JN387915.1 | 9 | 1237 | ||
| JN387916.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3646422 | Chr.21:14095581 - 14339672 on Build GRCh38 | Gain |
|
| nsv1056787 | Chr.21:14035183 - 14157970 on Build GRCh38 | Loss |
|
| nsv1058202 | Chr.21:13334446 - 14256347 on Build GRCh38 | Loss |
|
| nsv587043 | Chr.21:14129201 - 14178114 on Build GRCh38 | Loss |
|
| nsv1058072 | Chr.21:14000188 - 14195850 on Build GRCh38 | Gain |
|
| esv3893371 | Chr.21:14104569 - 14339246 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map