Assay Details
Target Gene Details
Entrez Gene ID: | 4719 |
Gene Name: | NADH:ubiquinone oxidoreductase core subunit S1 |
Gene Aliases: |
CI-75Kd, CI-75k, PRO1304 |
Location: |
Chr.2:206121971-206159519 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFS1 | NM_001199981.1 | 18 | 2240 | NP_001186910.1 |
| NM_001199982.1 | 16 | 2071 | NP_001186911.1 | |
| NM_001199983.1 | 18 | 2256 | NP_001186912.1 | |
| NM_001199984.1 | 19 | 2268 | NP_001186913.1 | |
| NM_005006.6 | 19 | 2348 | NP_004997.4 | |
| XM_017004188.1 | 19 | 2307 | XP_016859677.1 | |
| AK098395.1 | 14 | 1572 | ||
| AK295705.1 | 18 | 2146 | ||
| AK295966.1 | 15 | 1912 | ||
| AK295987.1 | 19 | 2268 | ||
| AK298320.1 | 16 | 1977 | ||
| AK300585.1 | 18 | 2160 | ||
| AW074155.1 | 1 | 496 | ||
| BC012068.1 | 7 | 854 | AAH12068.1 | |
| BC022368.1 | 19 | 2249 | AAH22368.1 | |
| BC030833.1 | 19 | 2252 | AAH30833.1 | |
| X61100.1 | 19 | 2211 | CAA43412.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893538 | Chr.2:206115621 - 206147759 on Build GRCh38 | Loss |
|
| esv3893539 | Chr.2:206123849 - 206133326 on Build GRCh38 | Loss |
|
| nsv821884 | Chr.2:206123804 - 206124254 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map