Assay Details
Target Gene Details
Entrez Gene ID: | 4025 |
Gene Name: | lactoperoxidase |
Gene Aliases: |
SPO |
Location: |
Chr.17:58238426-58268518 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LPO | NM_001160102.1 | NP_001153574.1 | ||
| NM_006151.2 | NP_006142.1 | |||
| NR_027647.1 | ||||
| XM_011524808.2 | XP_011523110.1 | |||
| AK131496.1 | 2 | 1105 | ||
| AK300653.1 | ||||
| AK303903.1 | ||||
| AK308378.1 | ||||
| AK310965.1 | ||||
| BC107166.1 | AAI07167.1 | |||
| BC107167.1 | AAI07168.1 | |||
| BX487327.1 | ||||
| BX648309.1 | ||||
| DB230339.1 | ||||
| EF579964.1 | ||||
| U39573.1 | AAC50717.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517229 | Chr.17:58176226 - 58373163 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map