Assay Details
Target Gene Details
Entrez Gene ID: | 4775 |
Gene Name: | nuclear factor of activated T-cells 3 |
Gene Aliases: |
NFAT4, NFATX |
Location: |
Chr.16:68085366-68229259 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFATC3 | NM_004555.3 | 2 | 824 | NP_004546.1 |
| NM_173163.2 | 2 | 824 | NP_775186.1 | |
| NM_173165.2 | 2 | 824 | NP_775188.1 | |
| AB209824.1 | BAD93061.1 | |||
| AK290370.1 | 2 | 564 | ||
| BC001050.2 | 2 | 542 | AAH01050.1 | |
| BU849493.1 | ||||
| EU887605.1 | 2 | 508 | ||
| EU887606.1 | 2 | 508 | ||
| EU887607.1 | 2 | 508 | ||
| EU887608.1 | 2 | 835 | ||
| EU887609.1 | 2 | 835 | ||
| EU887610.1 | 2 | 835 | ||
| EU887611.1 | 2 | 553 | ||
| EU887612.1 | 2 | 553 | ||
| EU887613.1 | 2 | 553 | ||
| EU887614.1 | 2 | 461 | ||
| EU887615.1 | 2 | 461 | ||
| EU887616.1 | 2 | 461 | ||
| EU887617.1 | 2 | 466 | ||
| EU887618.1 | 2 | 466 | ||
| EU887619.1 | 2 | 466 | ||
| EU887620.1 | 2 | 503 | ||
| EU887621.1 | 2 | 503 | ||
| EU887622.1 | 2 | 503 | ||
| EU887623.1 | 2 | 907 | ||
| EU887624.1 | 2 | 907 | ||
| EU887625.1 | 2 | 907 | ||
| L41067.1 | 2 | 718 | AAA79174.1 | |
| U14510.1 | 2 | 532 | AAA86308.1 | |
| U85428.1 | 2 | 532 | AAB46595.1 | |
| U85429.1 | 2 | 532 | AAB46596.1 | |
| U85430.1 | 2 | 532 | AAB46597.1 |
More Information
Set Membership: |
|

Genomic Map