Assay Details
Target Gene Details
Entrez Gene ID: | 165829 |
Gene Name: | G protein-coupled receptor 156 |
Gene Aliases: |
GABABL, PGR28 |
Location: |
Chr.3:120165481-120285122 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR156 | NM_001168271.1 | 9 | 2196 | NP_001161743.1 |
| NM_153002.2 | 9 | 2208 | NP_694547.2 | |
| XM_005247165.2 | 10 | 2485 | XP_005247222.1 | |
| XM_011512484.2 | 11 | 2558 | XP_011510786.1 | |
| XM_011512487.1 | 9 | 2110 | XP_011510789.1 | |
| XM_011512488.2 | 2 | 1177 | XP_011510790.1 | |
| XM_017005795.1 | 10 | 2521 | XP_016861284.1 | |
| XM_017005796.1 | 10 | 2530 | XP_016861285.1 | |
| XM_017005797.1 | 10 | 2473 | XP_016861286.1 | |
| AB209362.1 | 4 | 1372 | BAD92599.1 | |
| AF488739.1 | 9 | 2011 | AAN03795.1 | |
| BC113701.1 | 9 | 2031 | ||
| BC143606.1 | 9 | 2019 | ||
| BX115336.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1013788 | Chr.3:120147875 - 120193450 on Build GRCh38 | Gain |
|
| nsv829697 | Chr.3:120084009 - 120199588 on Build GRCh38 | Loss |
|
| nsv3967 | Chr.3:120138861 - 120184268 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map