Assay Details
Target Gene Details
Entrez Gene ID: | 7798 |
Gene Name: | leucine zipper protein 1 |
Gene Aliases: |
LUZP |
Location: |
Chr.1:23084017-23177808 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LUZP1 | NM_001142546.1 | 4 | 6523 | NP_001136018.1 |
| NM_033631.3 | 5 | 6589 | NP_361013.3 | |
| XM_011542090.2 | 6 | 7073 | XP_011540392.1 | |
| XM_011542091.2 | 5 | 7007 | XP_011540393.1 | |
| XM_017002252.1 | 7 | 7247 | XP_016857741.1 | |
| AK000297.1 | 1 | 1291 | ||
| AK025345.1 | 1 | 2478 | ||
| AL831896.1 | 1 | 2625 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6520 | Chr.1:23077876 - 23096553 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map