Assay Details
Target Gene Details
Entrez Gene ID: | 1643 |
Gene Name: | damage specific DNA binding protein 2 |
Gene Aliases: |
DDBB, UV-DDB2, XPE |
Location: |
Chr.11:47214942-47239218 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDB2 | NM_000107.2 | 10 | 1676 | NP_000098.1 |
NM_001300734.1 | 6 | 1109 | NP_001287663.1 | |
AA278953.1 | 1 | 195 | ||
AK091640.1 | 5 | 2011 | ||
BC000093.2 | 10 | 1504 | AAH00093.1 | |
U18300.1 | 10 | 1656 | AAB07897.1 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
