Assay Details
Target Gene Details
Entrez Gene ID: | 79411 |
Gene Name: | galactosidase beta 1 like |
Gene Aliases: |
- |
Location: |
Chr.2:219236598-219245429 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLB1L | NM_001286423.1 | 2 | 296 | NP_001273352.1 |
| NM_001286427.1 | 1 | 83 | NP_001273356.1 | |
| NM_024506.4 | 2 | 284 | NP_078782.3 | |
| XM_011511813.2 | XP_011510115.1 | |||
| XM_017004895.1 | 2 | 250 | XP_016860384.1 | |
| AK299404.1 | ||||
| AK300215.1 | 1 | 66 | ||
| AY358639.1 | 2 | 264 | AAQ89002.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834543 | Chr.2:219226398 - 219416430 on Build GRCh38 | Loss |
|
| esv3893555 | Chr.2:219172722 - 219273269 on Build GRCh38 | Loss |
|
| nsv508885 | Chr.2:219231225 - 219333602 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map