Assay Details
Target Gene Details
Entrez Gene ID: | 55812 |
Gene Name: | spermatogenesis associated 7 |
Gene Aliases: |
HEL-S-296, HSD-3.1, HSD3, LCA3 |
Location: |
Chr.14:88385638-88470350 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
