Assay Details
Target Gene Details
Entrez Gene ID: | 57658 |
Gene Name: | calcium binding and coiled-coil domain 1 |
Gene Aliases: |
Cocoa, PP13275, calphoglin |
Location: |
Chr.12:53711118-53727523 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CALCOCO1 | NM_001143682.1 | 14 | 2388 | NP_001137154.1 |
NM_020898.2 | 15 | 2643 | NP_065949.1 | |
NR_026554.1 | 14 | 2540 | ||
XM_011538601.1 | 16 | 2649 | XP_011536903.1 | |
XM_011538602.1 | 17 | 2782 | XP_011536904.1 | |
XM_011538603.1 | 17 | 2800 | XP_011536905.1 | |
XM_011538604.1 | 17 | 2756 | XP_011536906.1 | |
XM_017019707.1 | 16 | 2710 | XP_016875196.1 | |
XM_017019708.1 | 15 | 2777 | XP_016875197.1 | |
XM_017019709.1 | 14 | 2705 | XP_016875198.1 | |
AB040969.1 | 14 | 3518 | BAA96060.1 | |
AF370415.1 | 14 | 3526 | AAQ15251.1 | |
AK027881.1 | 15 | 2574 | BAB55428.1 | |
AK092788.1 | 14 | 2387 | ||
AK122773.1 | 14 | 2304 | ||
AK222646.1 | 10 | 1338 | ||
AL136895.1 | 15 | 2555 | CAB66829.1 | |
AY358397.1 | 15 | 2552 | AAQ88763.1 | |
AY563137.1 | 15 | 2550 | AAT68474.1 | |
BC003177.1 | 15 | 2550 | AAH03177.1 | |
BX538208.1 | 6 | 1121 | CAD98068.1 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
