Assay Details
Target Gene Details
Entrez Gene ID: | 933 |
Gene Name: | CD22 molecule |
Gene Aliases: |
SIGLEC-2, SIGLEC2 |
Location: |
Chr.19:35329166-35347361 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD22 | NM_001185099.1 | NP_001172028.1 | ||
| NM_001185100.1 | NP_001172029.1 | |||
| NM_001185101.1 | NP_001172030.1 | |||
| NM_001278417.1 | NP_001265346.1 | |||
| NM_001771.3 | NP_001762.2 | |||
| AK225625.1 | ||||
| AK225694.1 | ||||
| AK301177.1 | ||||
| BC109306.1 | AAI09307.1 | |||
| X52785.1 | CAA36988.1 | |||
| X59350.1 | CAA42006.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1067129 | Chr.19:35336133 - 35358408 on Build GRCh38 | Loss |
|
| nsv833810 | Chr.19:35291685 - 35481057 on Build GRCh38 | Gain |
|
| esv2758758 | Chr.19:35220175 - 35396211 on Build GRCh38 | Loss |
|
| nsv510765 | Chr.19:35217323 - 35399766 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map