Assay Details
Target Gene Details
Entrez Gene ID: | 1604 |
Gene Name: | CD55 molecule (Cromer blood group) |
Gene Aliases: |
CR, CROM, DAF, TC |
Location: |
Chr.1:207321472-207360966 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD55 | NM_000574.4 | 1 | 207 | NP_000565.1 |
| NM_001114752.2 | 1 | 207 | NP_001108224.1 | |
| NM_001300902.1 | 1 | 207 | NP_001287831.1 | |
| NM_001300903.1 | 1 | 207 | NP_001287832.1 | |
| NM_001300904.1 | 1 | 207 | NP_001287833.1 | |
| NR_125349.1 | 1 | 207 | ||
| XM_017000467.1 | XP_016855956.1 | |||
| AF052110.1 | 1 | 147 | ||
| AK298549.1 | ||||
| AK300621.1 | ||||
| AK312448.1 | ||||
| BC001288.1 | AAH01288.1 | |||
| BI545505.1 | 1 | 176 | ||
| DC386415.1 | 1 | 207 | ||
| M30142.1 | AAA52168.1 | |||
| M31516.1 | AAA52169.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1002014 | Chr.1:207223904 - 207359600 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map