Assay Details
Target Gene Details
Entrez Gene ID: | 1810 |
Gene Name: | down-regulator of transcription 1 |
Gene Aliases: |
NC2, NC2-BETA, NC2B |
Location: |
Chr.1:93345921-93362592 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DR1 | NM_001938.2 | NP_001929.1 | ||
| AK057541.1 | ||||
| AK223442.1 | BAD97162.1 | |||
| AL833729.1 | CAH56250.1 | |||
| BC002809.2 | AAH02809.1 | |||
| BC035507.2 | AAH35507.1 | |||
| BC068553.1 | AAH68553.1 |
More Information
Set Membership: |
|

Genomic Map