Assay Details
Target Gene Details
Entrez Gene ID: | 5351 |
Gene Name: | procollagen-lysine,2-oxoglutarate 5-dioxygenase 1 |
Gene Aliases: |
EDS6, LH, LH1, LLH, PLOD |
Location: |
Chr.1:11934667-11975542 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLOD1 | NM_000302.3 | 7 | 811 | NP_000293.2 |
| NM_001316320.1 | 8 | 952 | NP_001303249.1 | |
| AK294688.1 | 7 | 642 | ||
| AK295254.1 | ||||
| AK299150.1 | 8 | 885 | ||
| AK312243.1 | 7 | 744 | ||
| AK316285.1 | 8 | 888 | ||
| BC016657.2 | 7 | 715 | AAH16657.1 | |
| M98252.1 | 7 | 722 |
More Information
Set Membership: |
|

Genomic Map