Assay Details
Target Gene Details
Entrez Gene ID: | 387338 |
Gene Name: | NOP2/Sun RNA methyltransferase family member 4 |
Gene Aliases: |
SHTAP |
Location: |
Chr.1:46340177-46365152 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NSUN4 | NM_001256127.1 | 6 | 1770 | NP_001243056.1 |
| NM_001256128.1 | 6 | 1402 | NP_001243057.1 | |
| NM_199044.3 | 6 | 1825 | NP_950245.2 | |
| NR_045789.1 | 7 | 2000 | ||
| NR_045790.1 | 5 | 1670 | ||
| NR_045791.1 | 5 | 1488 | ||
| AK021577.1 | 6 | 1160 | BAB13847.1 | |
| AK057420.1 | 6 | 1761 | ||
| AK097524.1 | 6 | 1802 | ||
| AK128066.1 | 6 | 3541 | BAC87258.1 | |
| AK291741.1 | 6 | 1198 | ||
| AK292353.1 | 6 | 1376 | ||
| AK295003.1 | 7 | 1370 | ||
| BC014441.1 | 6 | 1246 | AAH14441.1 | |
| BX648592.1 | 5 | 1059 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829748 | Chr.1:46258836 - 46410450 on Build GRCh38 | Loss |
|
| esv2761743 | Chr.1:45874186 - 46561071 on Build GRCh38 | Gain |
|
| nsv546162 | Chr.1:46341925 - 46405089 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map