Assay Details
Target Gene Details
Entrez Gene ID: | 79594 |
Gene Name: | mitochondrial E3 ubiquitin protein ligase 1 |
Gene Aliases: |
C1orf166, GIDE, MAPL, MULAN, RNF218 |
Location: |
Chr.1:20499448-20508483 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MUL1 | NM_024544.2 | 4 | 1785 | NP_078820.2 |
| XM_011542137.2 | 4 | 2468 | XP_011540439.1 | |
| AB097015.1 | 4 | 1730 | BAC77368.1 | |
| AK022937.1 | 4 | 1771 | BAB14317.1 | |
| AL833889.1 | 4 | 1732 | CAD38745.1 | |
| BC010101.2 | 4 | 1692 | AAH10101.1 | |
| BC014010.1 | 4 | 1685 | AAH14010.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545731 | Chr.1:19989106 - 20776306 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map