Assay Details
Target Gene Details
Entrez Gene ID: | 90809 |
Gene Name: | transmembrane protein 55B |
Gene Aliases: |
C14orf9 |
Location: |
Chr.14:20457853-20461612 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM55B | NM_001100814.2 | NP_001094284.1 | ||
| NM_144568.3 | NP_653169.2 | |||
| AK314021.1 | ||||
| AL137727.1 | 2 | 2107 | CAB70896.1 | |
| BC002867.1 | AAH02867.2 | |||
| BC020947.1 | AAH20947.1 | |||
| BX161490.1 | CAD61939.1 | |||
| BX248025.1 | CAD62347.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv23438 | Chr.14:20332606 - 20789719 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map