Assay Details
Target Gene Details
Entrez Gene ID: | 540 |
Gene Name: | ATPase copper transporting beta |
Gene Aliases: |
PWD, WC1, WD, WND |
Location: |
Chr.13:51932669-52012130 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP7B | NM_000053.3 | 21 | 4988 | NP_000044.2 |
| NM_001005918.2 | 17 | 4367 | NP_001005918.1 | |
| NM_001243182.1 | 22 | 4655 | NP_001230111.1 | |
| XM_005266423.2 | 21 | 4893 | XP_005266480.1 | |
| XM_005266424.4 | 21 | 5514 | XP_005266481.1 | |
| XM_005266427.2 | 20 | 4816 | XP_005266484.1 | |
| XM_005266428.1 | 19 | 4798 | XP_005266485.1 | |
| XM_005266430.4 | 22 | 4996 | XP_005266487.1 | |
| XM_005266431.3 | 21 | 5048 | XP_005266488.1 | |
| XM_005266432.2 | 18 | 4564 | XP_005266489.1 | |
| XM_006719837.3 | 21 | 5142 | XP_006719900.1 | |
| XM_006719838.1 | 15 | 2783 | XP_006719901.1 | |
| XM_006719839.1 | 14 | 2600 | XP_006719902.1 | |
| XM_011535117.2 | 21 | 6069 | XP_011533419.1 | |
| XM_011535118.1 | 20 | 4915 | XP_011533420.1 | |
| XM_011535119.1 | 20 | 4867 | XP_011533421.1 | |
| XM_011535122.2 | 20 | 3741 | XP_011533424.1 | |
| XM_017020627.1 | 21 | 4892 | XP_016876116.1 | |
| XM_017020628.1 | 20 | 3737 | XP_016876117.1 | |
| AB209461.1 | 17 | 5577 | BAD92698.1 | |
| BX648261.1 | 14 | 4179 | ||
| L25591.1 | 16 | 3765 | AAA79211.1 | |
| U11700.1 | 21 | 4993 | AAA92667.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892343 | Chr.13:51418581 - 52060832 on Build GRCh38 | Gain |
|
| nsv561649 | Chr.13:51917057 - 51996592 on Build GRCh38 | Loss |
|
| dgv1652n100 | Chr.13:51747428 - 52069788 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map