Assay Details
Target Gene Details
Entrez Gene ID: | 5786 |
Gene Name: | protein tyrosine phosphatase, receptor type A |
Gene Aliases: |
HEPTP, HLPR, HPTPA, HPTPalpha, LRP, PTPA, PTPRL2, R-PTP-alpha, RPTPA |
Location: |
Chr.20:2864195-3038669 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPRA | NM_002836.3 | NP_002827.1 | ||
| M34668.1 | AAA36528.1 |
Target Gene Details
Entrez Gene ID: | 64601 |
Gene Name: | VPS16, CORVET/HOPS core subunit |
Gene Aliases: |
hVPS16 |
Location: |
Chr.20:2840697-2866732 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VPS16 | NM_022575.3 | NP_072097.2 | ||
| NM_080413.2 | NP_536338.1 | |||
| AF308801.1 | AAG34678.1 | |||
| AK055787.1 | BAB71013.1 | |||
| AL834401.1 | CAD39063.1 | |||
| BC012422.1 | AAH12422.1 | |||
| BC021291.2 | AAH21291.2 | |||
| BC064406.1 | AAH64406.1 | |||
| BC073959.1 | AAH73959.1 | |||
| BC126431.1 | ||||
| BM468030.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3645020 | Chr.20:2816405 - 2926274 on Build GRCh38 | Gain |
|
| nsv1057464 | Chr.20:2650623 - 2989686 on Build GRCh38 | Gain |
|
| nsv522009 | Chr.20:2745032 - 3030542 on Build GRCh38 | Gain |
|
| nsv833895 | Chr.20:2730164 - 2882236 on Build GRCh38 | Loss |
|
| esv2762056 | Chr.20:2740745 - 2944353 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map