Assay Details
Target Gene Details
Entrez Gene ID: | 80155 |
Gene Name: | N(alpha)-acetyltransferase 15, NatA auxiliary subunit |
Gene Aliases: |
Ga19, NARG1, NAT1P, NATH, TBDN, TBDN100 |
Location: |
Chr.4:139301467-139390781 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAA15 | NM_057175.3 | 1 | 177 | NP_476516.1 |
| XM_005263236.2 | 1 | 232 | XP_005263293.1 | |
| AF327722.1 | 1 | 200 | AAK15707.1 | |
| AJ314788.1 | 1 | 129 | CAC43228.1 | |
| AK023402.1 | 1 | 200 | BAB14562.1 | |
| AK304417.1 | ||||
| AK314626.1 | 1 | 232 | ||
| AY039242.1 | 1 | 202 | AAK68661.1 | |
| BC032642.1 | 1 | 202 | AAH32642.1 | |
| BC039818.1 | 1 | 192 | AAH39818.1 | |
| BC093928.1 | AAH93928.1 | |||
| BC104806.1 | AAI04807.1 |
Target Gene Details
Entrez Gene ID: | 4717 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit C1 |
Gene Aliases: |
KFYI |
Location: |
Chr.4:139289917-139302551 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFC1 | NM_001184987.1 | NP_001171916.1 | ||
| NM_001184988.1 | NP_001171917.1 | |||
| NM_001184989.1 | NP_001171918.1 | |||
| NM_001184990.1 | NP_001171919.1 | |||
| AK023115.1 | 1 | 774 | ||
| BM768825.1 | ||||
| BU677336.1 | ||||
| CB160001.1 | ||||
| CN347371.1 |
More Information
Set Membership: |
|

Genomic Map