Assay Details
Target Gene Details
Entrez Gene ID: | 28958 |
Gene Name: | cytochrome c oxidase assembly factor 3 |
Gene Aliases: |
CCDC56, HSPC009, MITRAC12 |
Location: |
Chr.17:42797618-42798725 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COA3 | NM_001040431.2 | 1 | 170 | NP_001035521.1 |
| AF070665.1 | 1 | 145 | AAD20971.1 | |
| AK225611.1 | 1 | 149 | ||
| AK290863.1 | 1 | 136 | ||
| AK295023.1 | 1 | 135 | ||
| AL524532.3 | 1 | 170 | ||
| AU122748.1 | 1 | 136 | ||
| BC002698.2 | 1 | 131 | AAH02698.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833450 | Chr.17:42660985 - 42825144 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map