Assay Details
Target Gene Details
Entrez Gene ID: | 1081 |
Gene Name: | glycoprotein hormones, alpha polypeptide |
Gene Aliases: |
CG-ALPHA, FSHA, GPHA1, GPHa, HCG, LHA, TSHA |
Location: |
Chr.6:87085498-87095147 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CGA | NM_000735.3 | NP_000726.1 | ||
| NM_001252383.1 | NP_001239312.1 | |||
| AK311972.1 | ||||
| BC010957.1 | AAH10957.1 | |||
| BC020782.1 | AAH20782.1 | |||
| BC055080.1 | AAH55080.1 | |||
| BG571584.1 | ||||
| BG619154.1 | ||||
| V00518.1 | CAA23777.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv604106 | Chr.6:85995537 - 87138584 on Build GRCh38 | Gain |
|
| esv2763585 | Chr.6:85938269 - 87211344 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map