Assay Details
Target Gene Details
Entrez Gene ID: | 89797 |
Gene Name: | neuron navigator 2 |
Gene Aliases: |
HELAD1, POMFIL2, RAINB1, STEERIN2, UNC53H2 |
Location: |
Chr.11:19345200-20121601 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
Target Gene Details
Entrez Gene ID: | 100874014 |
Gene Name: | NAV2 antisense RNA 2 |
Gene Aliases: |
- |
Location: |
Chr.11:20043775-20049303 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAV2-AS2 | NR_046672.1 | 2 | 572 | |
| BX112419.1 | 2 | 572 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv331e212 | Chr.11:20038899 - 20047215 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map