Assay Details
Target Gene Details
Entrez Gene ID: | 81873 |
Gene Name: | actin related protein 2/3 complex subunit 5 like |
Gene Aliases: |
ARC16-2 |
Location: |
Chr.9:124862118-124877733 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARPC5L | NM_030978.2 | 6 | 1929 | NP_112240.1 |
| XM_011519067.1 | 4 | 1063 | XP_011517369.1 | |
| AF087842.1 | 4 | 717 | AAP97155.1 | |
| AK023881.1 | 1 | 1686 | ||
| AK056174.1 | 6 | 1917 | ||
| BC000798.1 | 4 | 751 | AAH00798.1 | |
| BX537659.1 | 3 | 5924 |
More Information
Additional Information:
For this assay, SNP(s) [rs112734874] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map