Assay Details
Target Gene Details
Entrez Gene ID: | 64403 |
Gene Name: | cadherin 24 |
Gene Aliases: |
CDH11L |
Location: |
Chr.14:23047061-23057619 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDH24 | NM_022478.3 | NP_071923.2 | ||
| NM_144985.3 | NP_659422.2 | |||
| XM_011537089.1 | XP_011535391.1 | |||
| AK057922.1 | BAB71613.1 | |||
| AK289576.1 | ||||
| AK307746.1 | ||||
| AY260900.1 | AAP20590.1 | |||
| AY260901.1 | AAP20591.1 | |||
| AY358199.1 | AAQ88566.1 | |||
| BC110867.1 | AAI10868.1 | |||
| BX248750.1 | CAD66557.1 |
More Information
Set Membership: |
|

Genomic Map