Assay Details
Target Gene Details
Entrez Gene ID: | 84100 |
Gene Name: | ADP ribosylation factor like GTPase 6 |
Gene Aliases: |
BBS3, RP55 |
Location: |
Chr.3:97762875-97812585 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARL6 | NR_103511.1 | |||
| XM_006713783.3 | XP_006713846.1 | |||
| XM_017007311.1 | XP_016862800.1 | |||
| XM_017007312.1 | XP_016862801.1 | |||
| XM_017007313.1 | XP_016862802.1 | |||
| AK292958.1 | ||||
| AK310829.1 | ||||
| AK311281.1 | ||||
| AL136815.1 | CAB66749.1 | |||
| BC024239.2 | AAH24239.1 | |||
| BG722744.1 | ||||
| BM705017.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011526 | Chr.3:97483978 - 97905097 on Build GRCh38 | Gain |
|
| nsv520553 | Chr.3:95905623 - 98010484 on Build GRCh38 | Gain |
|
| nsv1013060 | Chr.3:96856275 - 98493673 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map