Assay Details
Target Gene Details
Entrez Gene ID: | 10677 |
Gene Name: | advillin |
Gene Aliases: |
ADVIL, DOC6, p92 |
Location: |
Chr.12:57797372-57818704 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AVIL | NM_006576.3 | 19 | 2416 | NP_006567.3 |
| XM_017018710.1 | 20 | 2482 | XP_016874199.1 | |
| XM_017018711.1 | 20 | 5228 | XP_016874200.1 | |
| XM_017018712.1 | 20 | 5171 | XP_016874201.1 | |
| XM_017018713.1 | 20 | 4831 | XP_016874202.1 | |
| XM_017018714.1 | 12 | 1631 | XP_016874203.1 | |
| XM_017018715.1 | 12 | 1505 | XP_016874204.1 | |
| AF041449.1 | 19 | 2416 | AAC25051.1 | |
| AK314362.1 | 19 | 2818 | ||
| BC111730.1 | 20 | 2520 | AAI11731.1 | |
| BX647344.1 | 9 | 2333 |
Target Gene Details
Entrez Gene ID: | 10102 |
Gene Name: | Ts translation elongation factor, mitochondrial |
Gene Aliases: |
EFTS, EFTSMT |
Location: |
Chr.12:57782745-57802856 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSFM | NM_001172697.1 | NP_001166168.1 | ||
| AK308981.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv559046 | Chr.12:57768302 - 58084815 on Build GRCh38 | Gain |
|
| nsv470297 | Chr.12:57739473 - 57808636 on Build GRCh38 | Gain |
|
| nsv510601 | Chr.12:57736515 - 57798142 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map