Assay Details
Target Gene Details
Entrez Gene ID: | 1027 |
Gene Name: | cyclin dependent kinase inhibitor 1B |
Gene Aliases: |
CDKN4, KIP1, MEN1B, MEN4, P27KIP1 |
Location: |
Chr.12:12717270-12722383 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDKN1B | NM_004064.4 | 1 | 547 | NP_004055.1 |
| AB451291.1 | ||||
| AB451423.1 | ||||
| AF247551.1 | AAF69497.1 | |||
| AJ616234.1 | CAE82383.1 | |||
| AK298335.1 | ||||
| AK312461.1 | 1 | 451 | ||
| AY004255.1 | 1 | 129 | AAF88142.1 | |
| BC001971.1 | 1 | 354 | AAH01971.1 | |
| BG701047.1 | 1 | 447 | ||
| BT019553.1 | AAV38360.1 | |||
| BT019554.1 | AAV38361.1 | |||
| CR457399.1 | CAG33680.1 | |||
| DA948043.1 | 1 | 449 | ||
| U10906.1 | AAA20240.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745567 | Chr.12:12246086 - 13092998 on Build GRCh38 | Deletion |
|
| nsv1046355 | Chr.12:11501836 - 12890185 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map