Assay Details
Target Gene Details
Entrez Gene ID: | 55668 |
Gene Name: | G-patch domain containing 2 like |
Gene Aliases: |
C14orf118 |
Location: |
Chr.14:76151885-76214473 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3581315 | Chr.14:76155402 - 76173245 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map