Assay Details
Target Gene Details
Entrez Gene ID: | 7738 |
Gene Name: | zinc finger protein 184 |
Gene Aliases: |
kr-ZNF3 |
Location: |
Chr.6:27450742-27473118 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF184 | NM_001318891.1 | NP_001305820.1 | ||
| NM_001318892.1 | NP_001305821.1 | |||
| NM_001318893.1 | NP_001305822.1 | |||
| NM_007149.2 | NP_009080.2 | |||
| XM_011514861.2 | XP_011513163.1 | |||
| AK123011.1 | 1 | 660 | ||
| AK131264.1 | BAD18442.1 | |||
| AK312804.1 | ||||
| BC022992.1 | AAH22992.1 | |||
| BX496218.1 | ||||
| CX870289.1 | ||||
| DB076907.1 | ||||
| DB090589.1 | ||||
| DB244087.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830617 | Chr.6:27456990 - 27630816 on Build GRCh38 | Loss |
|
| esv3570827 | Chr.6:27465036 - 27476385 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map