Assay Details
Target Gene Details
Entrez Gene ID: | 55924 |
Gene Name: | family with sequence similarity 212 member B |
Gene Aliases: |
C1orf183 |
Location: |
Chr.1:111722064-111755797 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM212B | NM_019099.4 | 2 | 313 | NP_061972.1 |
NM_198926.2 | 2 | 171 | NP_945120.1 | |
XM_011541783.2 | 4 | 409 | XP_011540085.1 | |
AK055667.1 | 2 | 289 | ||
AK293989.1 | 2 | 198 | ||
AK295012.1 | 2 | 252 | ||
AL137198.1 | 2 | 220 | CAB69908.1 | |
AL137199.1 | 2 | 171 | CAB69909.1 | |
BC031558.1 | 2 | 268 | AAH31558.1 | |
DA306915.1 | 2 | 313 |
