Assay Details
Target Gene Details
Entrez Gene ID: | 51138 |
Gene Name: | COP9 signalosome subunit 4 |
Gene Aliases: |
CSN4 |
Location: |
Chr.4:83035086-83075818 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COPS4 | NM_001258006.1 | 1 | 157 | NP_001244935.1 |
| NM_016129.2 | 1 | 157 | NP_057213.2 | |
| XM_011532019.1 | 1 | 153 | XP_011530321.1 | |
| AF100757.1 | AAD43021.1 | |||
| AK001148.1 | 1 | 153 | ||
| AK001210.1 | BAA91555.1 | |||
| AK024005.1 | 1 | 42 | ||
| AK094238.1 | 1 | 796 | ||
| AK223178.1 | BAD96898.1 | |||
| BC004302.1 | AAH04302.1 | |||
| BC009292.2 | AAH09292.1 | |||
| BC093007.1 | 1 | 42 | AAH93007.1 | |
| CD690475.1 | 1 | 188 | ||
| KU178633.1 | ||||
| KU178634.1 |
More Information
Set Membership: |
|

Genomic Map