Assay Details
Target Gene Details
Entrez Gene ID: | 1831 |
Gene Name: | TSC22 domain family member 3 |
Gene Aliases: |
DIP, DSIPI, GILZ, TSC-22R |
Location: |
Chr.X:107713221-107777329 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TSC22D3 | NM_001015881.1 | 3 | 618 | NP_001015881.1 |
NM_001318468.1 | 4 | 1086 | NP_001305397.1 | |
NM_001318470.1 | 4 | 1072 | NP_001305399.1 | |
NM_004089.3 | 3 | 823 | NP_004080.2 | |
NM_198057.2 | 3 | 1139 | NP_932174.1 | |
XM_005262099.1 | 5 | 1100 | XP_005262156.1 | |
XM_005262100.1 | 4 | 975 | XP_005262157.1 | |
XM_005262102.1 | 4 | 968 | XP_005262159.1 | |
XM_005262103.3 | 4 | 1078 | XP_005262160.1 | |
XM_011530884.1 | 4 | 1174 | XP_011529186.1 | |
XM_017029335.1 | 4 | 910 | XP_016884824.1 | |
AB025432.1 | 3 | 806 | BAB18680.1 | |
AF153603.1 | 3 | 801 | AAD41085.1 | |
AF183393.1 | 3 | 809 | AAD56234.1 | |
AF228339.1 | 3 | 813 | AAG12456.1 | |
AK092645.1 | 4 | 1025 | BAC03934.1 | |
AK092669.1 | 3 | 1111 | ||
AK127938.1 | 3 | 763 | ||
AK300535.1 | 4 | 950 | ||
AL110191.1 | 3 | 822 | CAB53669.1 | |
AY007119.1 | 3 | 722 | ||
BC018148.1 | 3 | 505 | ||
BC072446.1 | 3 | 817 | AAH72446.1 | |
BX647854.1 | 4 | 1072 | ||
CR933650.1 | 3 | 825 | CAI45951.1 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
