Assay Details
Target Gene Details
Entrez Gene ID: | 101928034 |
Gene Name: | uncharacterized LOC101928034 |
Gene Aliases: |
- |
Location: |
Chr.1:153545742-153551769 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928034 | NR_125947.1 | |||
| AK307246.1 | ||||
| DA480135.1 |
Target Gene Details
Entrez Gene ID: | 6274 |
Gene Name: | S100 calcium binding protein A3 |
Gene Aliases: |
S100E |
Location: |
Chr.1:153547333-153549258 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100A3 | NM_002960.1 | 3 | 328 | NP_002951.1 |
| BC012893.1 | 3 | 312 | AAH12893.1 | |
| BT006955.1 | AAP35601.1 | |||
| CR542163.1 | CAG46960.1 | |||
| CR542185.1 | CAG46982.1 | |||
| Z18948.1 | 3 | 328 | CAA79471.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831603 | Chr.1:153433366 - 153603667 on Build GRCh38 | Gain |
|
| nsv1002705 | Chr.1:153335376 - 153553113 on Build GRCh38 | Gain |
|
| esv3587591 | Chr.1:153528630 - 153570193 on Build GRCh38 | Gain |
|
| esv3587584 | Chr.1:153466126 - 153598991 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map