Assay Details
Target Gene Details
Entrez Gene ID: | 112 |
Gene Name: | adenylate cyclase 6 |
Gene Aliases: |
AC6, LCCS8 |
Location: |
Chr.12:48766191-48789096 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADCY6 | NM_015270.4 | NP_056085.1 | ||
| XM_006719210.3 | XP_006719273.1 | |||
| XM_017018743.1 | XP_016874232.1 | |||
| AB007882.2 | ||||
| AF250226.1 | AAF82478.1 | |||
| BX537664.1 |
More Information
Set Membership: |
|

Genomic Map