Assay Details
Target Gene Details
Entrez Gene ID: | 8202 |
Gene Name: | nuclear receptor coactivator 3 |
Gene Aliases: |
ACTR, AIB-1, AIB1, CAGH16, CTG26, KAT13B, RAC3, SRC-3, SRC3, TNRC14, TNRC16, TRAM-1, bHLHe42, pCIP |
Location: |
Chr.20:47501857-47656877 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCOA3 | NM_001174087.1 | 23 | 6101 | NP_001167558.1 |
| NM_001174088.1 | 23 | 6077 | NP_001167559.1 | |
| NM_006534.3 | 23 | 6092 | NP_006525.2 | |
| NM_181659.2 | 23 | 6104 | NP_858045.1 | |
| AF012108.1 | 23 | 6031 | AAC51677.1 | |
| AF036892.1 | 23 | 5990 | AAB92368.1 | |
| BC119001.1 | 23 | 5980 | ||
| BC122547.1 | 23 | 5976 |
Target Gene Details
Entrez Gene ID: | 55959 |
Gene Name: | sulfatase 2 |
Gene Aliases: |
HSULF-2 |
Location: |
Chr.20:47654584-47786629 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SULF2 | XM_005260458.2 | 21 | 6235 | XP_005260515.1 |
| XM_017027955.1 | 21 | 6307 | XP_016883444.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758799 | Chr.20:47639666 - 47895335 on Build GRCh38 | Gain |
|
| nsv1066907 | Chr.20:47310827 - 47736827 on Build GRCh38 | Gain |
|
| nsv833995 | Chr.20:47550035 - 47694664 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map