Assay Details
Target Gene Details
Entrez Gene ID: | 4256 |
Gene Name: | matrix Gla protein |
Gene Aliases: |
GIG36, MGLAP, NTI |
Location: |
Chr.12:14881181-14885919 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGP | NM_000900.3 | NP_000891.2 | ||
| NM_001190839.1 | NP_001177768.1 | |||
| AK312029.1 | ||||
| AW997681.1 | ||||
| AY542304.1 | AAT08173.1 | |||
| BC005272.1 | AAH05272.1 | |||
| BC070314.1 | AAH70314.1 | |||
| BC093078.1 | AAH93078.1 | |||
| BT006733.1 | AAP35379.1 | |||
| CB069816.1 | ||||
| CB108937.1 | ||||
| CR450358.1 | CAG29354.1 | |||
| M58549.1 | AAB53766.1 | |||
| X07362.1 | CAA30287.1 | |||
| X53331.1 | CAA37418.1 |
More Information
Set Membership: |
|

Genomic Map