Assay Details
Target Gene Details
Entrez Gene ID: | 58477 |
Gene Name: | SRP receptor beta subunit |
Gene Aliases: |
APMCF1 |
Location: |
Chr.3:133784033-133821492 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRPRB | NM_021203.3 | 2 | 386 | NP_067026.3 |
| AF086914.1 | 1 | 124 | AAP97144.1 | |
| AF141882.3 | 1 | 117 | AAD34888.3 | |
| AK027525.1 | 1 | 117 | BAB55176.1 | |
| AK075531.1 | 1 | 155 | BAC11675.1 | |
| BC063001.1 | 1 | 129 | AAH63001.1 | |
| BC065299.1 | 1 | 101 | AAH65299.1 | |
| DB448771.1 | 2 | 386 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829729 | Chr.3:133726027 - 133901543 on Build GRCh38 | Loss |
|
| nsv4018 | Chr.3:133767023 - 133811824 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map