Assay Details
Target Gene Details
Entrez Gene ID: | 26256 |
Gene Name: | calcium binding tyrosine phosphorylation regulated |
Gene Aliases: |
CABYRa, CABYRc, CABYRc/d, CABYRe, CBP86, CT88, FSP-2, FSP2 |
Location: |
Chr.18:24138956-24161600 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CABYR | NM_001308231.1 | 2 | 960 | NP_001295160.1 |
NM_012189.3 | 4 | 1183 | NP_036321.2 | |
NM_138643.2 | NP_619584.1 | |||
NM_138644.2 | NP_619585.1 | |||
NM_153768.2 | 3 | 1129 | NP_722452.1 | |
NM_153769.2 | NP_722453.1 | |||
NM_153770.2 | NP_722454.1 | |||
XM_005258247.2 | XP_005258304.1 | |||
XM_017025706.1 | 4 | 1183 | XP_016881195.1 | |
AF088868.1 | 4 | 1044 | AAC35373.1 | |
AF295037.1 | AAG17889.1 | |||
AF295038.1 | AAG17890.1 | |||
AF295039.1 | AAG17891.1 | |||
AF329634.1 | 3 | 990 | AAL56051.1 | |
AK225858.1 | ||||
AK301889.1 | 2 | 855 | ||
AK313243.1 | 3 | 1068 | ||
AY007205.1 | 1 | 393 | AAG01891.1 | |
BC011996.1 | AAH11996.1 | |||
HM594163.1 | 1 | 480 | ||
HM594164.1 | 3 | 996 | ||
HM594165.1 | 3 | 996 | ||
HM594166.1 | 3 | 996 | ||
HM594167.1 | ||||
HM594168.1 | ||||
HM594169.1 | ||||
HQ659768.1 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
