Assay Details
Target Gene Details
Entrez Gene ID: | 5268 |
Gene Name: | serpin family B member 5 |
Gene Aliases: |
PI5, maspin |
Location: |
Chr.18:63476911-63505085 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINB5 | NM_002639.4 | NP_002630.2 | ||
| AK301667.1 | ||||
| AK312765.1 | ||||
| AL602439.1 | ||||
| BC020713.1 | AAH20713.1 | |||
| U04313.1 | AAA18957.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066552 | Chr.18:63147299 - 64608176 on Build GRCh38 | Gain |
|
| nsv131597 | Chr.18:63473298 - 63482700 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75113857] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map